- Antibody Type:Primary
- Antigen Name:interferon, gamma
- Antigen Symbol:IFNG
- Clonality:Monoclonal
- Clone:IFNG/466
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P01579
- Antigen Synonyms:IFNG
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Human recombinant protein was used as the immunogen for the IFN gamma antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-392
Recognizes a protein of 20-25kDa, identified as human interferon. This mAb is specific to human IFNg and recognizes both recombinant and native human IFNg. T lymphocytes and NK cells mainly produce IFNg. It is a pleiotropic cytokine involved in the regulation of nearly all phases of immune and inflammatory responses, including the activation, growth and differentiation of T cell, B cells, macrophages, NK cells and other cell types such as endothelial cells and fibroblasts. It has weak anti-viral and anti-proliferative activity, and potentiates the antiviral and anti-tumor effects of IFNa/b (type I interferon).
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 1-2 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 min at RT Optimal dilution of the IFN gamma antibody should be determined by the researcher.
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: IFNG/466
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human