Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-FUT4 Mouse Monoclonal Antibody [clone: Leu-M1 or MMA]
Anti-FUT4 Mouse Monoclonal Antibody [clone: Leu-M1 or MMA]
Catalog # 76194-374
Supplier:  Prosci
Anti-FUT4 Mouse Monoclonal Antibody [clone: Leu-M1 or MMA]
Catalog # 76194-374
Supplier:  Prosci
Supplier Number:  33-568

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific)
  • Antigen Symbol:
    FUT4
  • Clonality:
    Monoclonal
  • Clone:
    Leu-M1 or MMA
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgM kappa
  • Reactivity:
    Human
  • Size:
    100 ug
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P22083
  • Antigen Synonyms:
    FUT4
  • Storage Buffer:
    PBS with 0.1 mg/ml BSA and 0.05% sodium azide
  • Storage Temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0.2 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    The histiocytic cell line U937 was used as the immunogen for this CD15 antibody.
  • Purification:
    Protein G affinity chromatography
  • Cat. No.:
    76194-374

Specifications

About this item

CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin s disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the CD15 antibody for each application should be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: Leu-M1 or MMA
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human