Specifications
- Antibody Type:Primary
- Antigen Symbol:IPO38 Antibody [SPM260]
- Clonality:Monoclonal
- Clone:SPM260
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human,Rat,Mouse
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Spleen cells from a patient with hairy cell leukemia were used as the immunogen for the IPO38 antibody.
- Purification:PEG precipitation
- Cat. No.:76194-340
Specifications
About this item
Recognizes a protein of 14-16kDa, which is a novel nuclear antigen of proliferating cells. IPO-38 antigen is present in the nuclei of proliferating cells such as Hodgkin s disease and non-Hodgkin s lymphomas, different forms of leukemias, breast and colorectal carcinomas, and PHA-stimulated lymphocytes. It is not expressed in the cells of non-stimulated lymphocytes and granulocytes. IPO-38 can be a useful marker of cell proliferation during monitoring of tumor progression.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the IPO38 antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 min.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: IPO38 Antibody [SPM260]
Clonality: Monoclonal
Clone: SPM260
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human, Mouse, Rat