Specifications
- Antibody Type:Primary
- Antigen Name:Major capsid protein L1
- Antigen Symbol:L1
- Clonality:Monoclonal
- Clone:CamVir-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:HPV-16
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P03101
- Antigen Synonyms:L1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Human papilloma virus type 16, major capsid protein L1, was used as the immunogen for the L1 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-324
Specifications
About this item
Reacts with a protein of 57kDa, identified as the L1 protein of human papilloma virus type 16 (HPV-16). Forms an icosahedral capsid with a T=7 symmetry and a 50 nm diameter. The capsid is composed of 72 pentamers linked to each other by disulfide bonds and associated with L2 proteins. Binds to heparan sulfate proteoglycans on the basement membrane to provide initial virion attachment to target cells. Basement membrane is exposed only after epithelium trauma. Additionally, the alpha6 integrin complexed with either beta1 or beta4 integrin has been proposed to act as a coreceptor recognized by L1. Once attached, integrin complexed with beta4 integrin has been proposed to act as a coreceptor recognized by L1. Once attached, the virion enters the host cell via clathrin-mediated endocytosis and the genomic DNA is released to the host nucleus. The virion assembly takes place within the cell nucleus. Encapsulates the genomic DNA together with protein L2. [UniProt]
The antibody reacts very strongly with formalin-fixed, paraffin-embedded tissues containing HPV-16 or -33; very weak reactions were occasionally observed with biopsy specimens or smears containing HPV-6 or HPV-11. It cross-reacts with HPV37.
Western blot: 1-2 ug/ml ELISA: assay dependent Immunofluorescence: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 min at RT (1) Optimal dilution of the L1 antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 min.
Type: Primary
Antigen: L1
Clonality: Monoclonal
Clone: CamVir-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: HPV-16