Specifications
- Antibody Type:Primary
- Antigen Name:heat shock protein 90kDa beta (Grp94), member 1
- Antigen Symbol:HSP90B1
- Clonality:Monoclonal
- Clone:HSP90B1/1192
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P14625
- Antigen Synonyms:HSP90B1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant full-length human protein was used as the immunogen for the GRP94 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-226
Specifications
About this item
Recognizes a protein of 94kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This mAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grps are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 0.5-1 ug/ml Western blot: 0.5-1 ug/ml Immunohistology (FFPE): 0.5-1 ug/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the GRP94 antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 min.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: HSP90B1/1192
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Human