Specifications
- Antibody Type:Primary
- Antigen Name:Trefoil factor 1
- Antigen Symbol:TFF1
- Clonality:Monoclonal
- Clone:SPM313
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Cynomolgus monkey,Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P04155
- Antigen Synonyms:TFF1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Amino acids CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the human protein were used as the immunogen for the Estrogen Inducible Protein pS2 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-208
Specifications
About this item
It recognizes a polypeptide of 6.5kDa, identified as pS2 estrogen-regulated protein. Its epitope is localized between aa57-84 of human pS2 protein. pS2 is a trefoil peptide. Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. These peptides contain three intra-chain disulfide bonds, forming the trefoil motif, or P-domain. pS2 is known to form dimers and this dimerization is thought to play a role in its protective and healing properties. About 60% of breast carcinomas are positive for pS2. Staining is cytoplasmic, often with localization to the Golgi apparatus. pS2 is shown to be localized in normal stomach mucosa, gastric fluid, goblet cells in the colon and small intestine, and in ulcerations of the gastrointestinal tract. Several studies have shown that pS2 is primarily expressed in estrogen receptor-positive breast tumors and it may define a subset of estrogen-dependent tumors that displays an increased likelihood of response to endocrine therapy.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the Estrogen Inducible Protein pS2 antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 min.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: TFF1
Clonality: Monoclonal
Clone: SPM313
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Cynomolgus Monkey