- Antibody Type:Primary
- Antigen Name:carbamoyl-phosphate synthase 1, mitochondrial
- Antigen Symbol:CPS1
- Clonality:Monoclonal
- Clone:CPS1/1022
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Dog
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P31327
- Antigen Synonyms:CPS1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human protein was used as the immunogen for the CPS1 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-832
This mAb recognizes a protein of 165kDa, identified as carbamoyl phosphate synthetase 1 (CPS1). This mitochondrial enzyme catalyzes synthesis of carbamoyl phosphate from ammonia and bicarbonate. This reaction is the first committed step of the urea cycle, which is important in the removal of excess urea from cells. Deficiency of CPS1 is an autosomal recessive disorder that causes hyperammonemia. CPS1 is a hepatocyte specific protein that localizes to the mitochondria of hepatocytes. It is a sensitive marker for distinguishing hepatocellular carcinomas (HCC) from other metastatic carcinomas as well as cholangio-carcinomas. HCC s occur primarily in the stomach, but they are also found in many other organs. CPS1 may also be a useful marker for intestinal metaplasia. Reportedly, strong expression of CPS1 correlates with smaller tumor size and longer patient survival. Occasionally, CPS1 is also found in gastric carcinomas as well as in a few other non-hepatic tumors.
Immunofluorescence: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.25-0.5 ug/ml for 30 min at RT (1) Prediluted format : incubate for 30 min at RT (2) Optimal dilution of the CPS1 antibody should be determined by the researcher.
1.Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 10-20 min followed by cooling at RT for 20 min
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CPS1
Clonality: Monoclonal
Clone: CPS1/1022
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog