- Antibody Type:Primary
- Antigen Name:protein tyrosine phosphatase, receptor type, C
- Antigen Symbol:PTPRC
- Clonality:Monoclonal
- Clone:SPM125
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Common marmoset,Chimpanzee,Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P08575
- Antigen Synonyms:PTPRC
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Cultured human T-cells from an IL-2-dependent T-cell line (CA1) were used as immunogen for this anti-CD45RO antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76193-904
Recognizes a 180-185kDa protein, identified as isoform of leukocyte common antigen (CD45RO). This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas.
Flow Cytometry: 0.5-1 ug/million cells Immunofluorescence: 0.5-1 ug/ml Immunohistology (FFPE): 0.25-0.5 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) Due to variation in protocol and secondary antibody used, the anti-CD45RO antibody may need to be titered for optimal performance.
1. FFPE staining requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: SPM125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Chimpanzee, Common Marmoset