- Antibody Type:Primary
- Antigen Name:Cytomegalovirus Antibody [CMV100]
- Clonality:Monoclonal
- Clone:CMV100
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Cytomegalovirus
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Tested Applications:Immunocytochemistry
- Purification:Protein G purified CMV antibody
- Cat. No.:76195-698
Ten to forty percent of the patients with acquired immunodeficiency syndrome (AIDS) develop Cytomegalovirus infections. In some patients with AIDS, the virus is detected in the bronchoalveolar lavage fluid (BALF), urine, and other specimens, even when there are no symptoms of the disease. An indicator of active infection is needed to facilitate the diagnosis of cytomgalovirus disease in patients with AIDS or HIV infection. Cytomegalovirus was detected in the leukocytes of both the peripheral blood and BALF during the early phase of the disease.
ELISA: order BSA-free CMV antibody format for coating Immunocytochemistry (Acetone or paraformaldehyde fixed) 0.5-1 ug/ml for 30 min IF: 0.5-1 ug/ml The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the CMV antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: Cytomegalovirus Antibody [CMV100]
Clonality: Monoclonal
Clone: CMV100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Cytomegalovirus