To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:keratin 10
- Antigen Symbol:KRT10
- Clonality:Monoclonal
- Clone:AE1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Turtle,Rabbit,Monkey,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P13645
- Antigen Synonyms:KRT10
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Solubilized keratin extract from human stratum corneum
- Purification:Protein G purified
- Cat. No.:76195-694
Specifications
About this item
Cytokeratins, also called keratins or simply CKs, are a family of fibrous structural proteins and a major structural component in the outer layer of human skin, as well as hair and nails. There are 20 human epithelial keratins which can be divided into two subfamiles: acidic and basic (more recent human genone sequencing has identified an additional 20 members). The formation of cytokeratin intermediate filaments requires the pairing of at least one acidic and one basic subfamily member. Members of the same keratin subfamily share extended sequence homology while members of different subfamilies show only limited sequence homology.
This antibody is specific for the 56.5kDa (CK10), 50kDa (CK14), 50kDa (CK15), 48kDa (CK16), and 40kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. Many studies have shown the usefulness of CKs as markers in cancer research and tumor diagnosis, as epithelial tumors generally maintain the same CK expression patterns as their counterpart normal tissue. Clone AE1 is commonly used with clone AE3 as a pan cytokeratin antibody cocktail (Cat No V2330).
Flow Cytometry: 0.5-1 ug/million cells IF: 1-2 ug/ml WB: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 min at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Acidic Cytokeratin antibody AE1 to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: KRT10
Clonality: Monoclonal
Clone: AE1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Bovine, Dog, Rabbit, Mouse, Rat, Chicken, Turtle