Specifications
- Antibody Type:Primary
- Antigen Name:anoctamin 1, calcium activated chloride channel
- Antigen Symbol:ANO1
- Clonality:Monoclonal
- Clone:DG1/447
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q5XXA6
- Antigen Synonyms:ANO1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human TMEM16A/DOG1 protein was used as the immunogen for this TMEM16A antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-682
Specifications
About this item
Expression of TMEM16A (DOG1) protein is elevated in the gastrointestinal stromal tumors (GIST’s), c-kit signaling-driven mesenchymal tumors of the GI tract. It is rarely expressed in other soft tissue tumors, which, due to appearance, may be difficult to diagnose. Immunoreactivity for TMEM16A has been reported in 97.8 percent of scorable GIST’s, including all c-kit negative GIST’s. Overexpression has been suggested to aid in the identification of GISTs, including Platelet-Derived Growth Factor Receptor Alpha (PDGFR alpha) mutants that fail to express c-kit antigen. The overall sensitivity of TMEM16A and c-kit in GIST’s is nearly identical: 94.4% vs. 94.7%.
Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the TMEM16A antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: ANO1
Clonality: Monoclonal
Clone: DG1/447
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human