Specifications
- Antibody Type:Primary
- Antigen Name:Metalloproteinase inhibitor 3
- Antigen Symbol:TIMP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Horse,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P35625
- Antigen Synonyms:TIMP3
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Amino acids 175-211 of human TIMP3 Ab was used as the immunogen for this antibody (100% homologous in cow, dog and horse, and 94% homologous in mouse and rat).
- Purification:Protein A affinity chromatography
- Cat. No.:76193-882
Specifications
About this item
TIMP3 (tissue inhibitor of metalloproteinases 3), along with family members TIMP1, TIMP2, and TIMP4, are inhibitors of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix (ECM). An imbalance between MMPs and the associated TIMPs may play a significant role in the invasive phenotype of malignant tumors. TIMP’s inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells. TIMP3 may be involved in regulating trophoblastic invasion of the uterus as well as in regulating remodeling of the extracellular matrix during the folding of epithelia, and in the formation, branching and expansion of epithelial tubes.
Flow Cytometry: use antibody at 0.5-1 ug/million cells IF: 1-2 ug/ml WB: 0.5-1 ug/ml IP: 1-2ug/500ug protein lysate Immunohistochemistry (FFPE): 1-2 ug/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2)
The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the TIMP3 antibody to be titered up or down for optimal performance.1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: TIMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Horse, Bovine, Dog