- Antibody Type:Primary
- Antigen Name:solute carrier family 3 (amino acid transporter heavy chain), member 2
- Antigen Symbol:SLC3A2
- Clonality:Monoclonal
- Clone:UM7F8
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Neutralisation:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P08195
- Antigen Synonyms:SLC3A2
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:The Molt-13 T cell line was used as the immunogen for this CD98 antibody.
- Tested Applications:Functional
- Purification:Protein G purified CD98 antibody
- Cat. No.:76195-628
CD98 exits as a heterodimer containing a disulphide-linked glycosylated heavy chain and a non-glycosylated light chain. It is a member of the solute carrier family and encodes a cell surface, transmembrane protein. The protein exists as the heavy chain of a heterodimer, covalently bound through disulfide bonds to one of several possible light chains. The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids. Alternatively spliced transcript variants, encoding different isoforms, have been characterized.
Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1.0 ug/ml Functional Activity: Comitogenic with soluble anti-CD2 and immobilized anti-CD3 mAbs The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the CD98 antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: UM7F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human