Specifications
- Antibody Type:Primary
- Antigen Name:keratin 10
- Antigen Symbol:KRT10
- Clonality:Monoclonal
- Clone:LH2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P13645
- Antigen Synonyms:KRT10
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Skin extract from a human psoriasis patient was used as the immunogen for this Cytokeratin 10 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-556
Specifications
About this item
This antibody recognizes a protein of 56.5kDa, identified as cytokeratin 10 (CK10). cytokeratin 10 is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of cytokeratin 10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. However, cytokeratin 10 is rarely detected in early stages of vulvar squamous carcinomas (tumors less than 2 cm, clinical stage I) regardless of the tumor grade. In larger and more advanced tumors (greater than 2 cm, clinical stages II and III), cytokeratin 10 is detected very frequently. Expression of cytokeratin 10 is related to maturation of malignant keratinocytes, being preferentially detected in more-differentiated parts.
Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 1mM EDTA, pH 8-9, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: KRT10
Clonality: Monoclonal
Clone: LH2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse