Specifications
- Antibody Type:Primary
- Antigen Name:KIT proto-oncogene receptor tyrosine kinase
- Antigen Symbol:CD117
- Clonality:Monoclonal
- Clone:C117/370
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P10721
- Antigen Synonyms:CD117
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human protein was used as the immunogen for this CD117 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-546
Specifications
About this item
This antibody recognizes a protein of 145kDa, identified as CD117, or p145kit. It is found on a wide variety of tumor cells including follicular and papillary carcinoma of thyroid, adenocarcinomas from endometrium, lung, ovary, pancreas, and breast as well as malignant melanoma, endodermal sinus tumor, and small cell carcinoma. However, CD117 antibody has been particularly useful in differentiating gastrointestinal stromal tumors from Kaposi’s sarcoma, tumors of smooth muscle origin, fibromatosis, and neural tumors of the GI tract. CD117 antibody is also useful in recognizing myeloblasts in bone marrow biopsy and clot section.
IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Flow Cytometry: 0.5-1 ug/million cells in 0.1ml IF: 0.5-1 ug/ml The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the CD117 antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
Type: Primary
Antigen: CD117
Clonality: Monoclonal
Clone: C117/370
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human