Specifications
- Antibody Type:Primary
- Antigen Name:erb-b2 receptor tyrosine kinase 2
- Antigen Symbol:ERBB2
- Clonality:Monoclonal
- Clone:HRB2/451
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P04626
- Antigen Synonyms:ERBB2
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human protein was used as the immunogen for this HER2 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-506
Specifications
About this item
This mAb is specific to HER2 / ErbB2 and shows minimal cross-reaction with other members of the family. HER2 is a member of the EGFR family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. HER2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml IF: 1-2 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 min at Room Temp (1) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the mAb to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: HRB2/451
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human