- Antibody Type:Primary
- Antigen Name:Cell division cycle protein 20 homolog
- Antigen Symbol:CDC20
- Clonality:Monoclonal
- Clone:AR12
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q12834
- Antigen Synonyms:CDC20
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Denatured recombinant human protein was used as the immunogen for this Cdc20 antibody.
- Purification:Protein G purified antibody
- Cat. No.:76195-474
Cyclins, regulatory subunits which associate with kinases, control many of the important steps in cell cycle progression. The Cdc2 kinase (p34Cdc2) exhibits kinase activity in vitro and exists in a complex with both cyclin B and a protein homologous to p13SUC1. Cdc2 kinase is the active subunit of the M phase promoting factor (MPF) and the M phase-specific Histone H1 kinase. The p34Cdc2/cyclin B complex is required for the G2 to M transition. An additional cell cycle-dependent protein kinase, termed p55cdc, exhibits a high degree of homology with the S. cerevisiae proteins Cdc20 and Cdc4. The p55cdc transcript is readily detectable in a variety of cultured cell lines in growth phase, but disappears when cell growth is chemically arrested.
Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Cdc20 antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate Buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CDC20
Clonality: Monoclonal
Clone: AR12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human