Specifications
- Antibody Type:Primary
- Antigen Name:CD79a molecule, immunoglobulin-associated alpha
- Antigen Symbol:CD79A
- Clonality:Monoclonal
- Clone:JCB117
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P11912
- Antigen Synonyms:CD79A
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Amino acids 202-216 (GTYQDVGSLNIADVQ) were used as the immunogen for this CD79a antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-470
Specifications
About this item
A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells. This complex of mb-1 and B29 polypeptides and immunoglobulin constitute the B-cell Ag receptor. CD79a first appears at pre B-cell stage, early in maturation, and persists until the plasma cell stage where it is found as an intracellular component. It is found in the majority of acute leukemias of B-cell precursors, lines and lymphomas, and in some myelomas. It is not present in myeloid or Tcell lines. CD79a antibody is generally used to complement CD20 mAb, especially for mature B-cell lymphomas after treatment with Rituximab (anti-CD20). This CD79a antibody will stain many of the same lymphomas as CD20 mAb, but also is more likely to stain B-lymphoblastic lymphoma/leukemia than is CD20 mAb. CD79a antibody also stains more cases of plasma cell myeloma and occasionally some types of endothelial cells as well.
Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the CD79a antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: JCB117
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human