- Antibody Type:Primary
- Antigen Name:CD44 molecule (Indian blood group)
- Antigen Symbol:CD44
- Clonality:Monoclonal
- Clone:156-3C11
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Baboon,Green monkey,Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P16070
- Antigen Synonyms:CD44
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Stimulated human leukocytes were used as the immunogen for this CD44 / HCAM antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-458
This antibody recognizes a cell surface glycoprotein of 80-95kDa, called CD44, or HCAM, on lymphocytes, monocytes, and granulocytes (Leucocyte Typing Workshop V). Its epitope is resistant to digestion by trypsin and chymotrypsin. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95kDa or hematopoietic variant (CD44-s). Higher molecular weight isoforms are described in epithelial cells (CD44-v), which are believed to function in intercellular adhesion and stromal binding. HCAM antibody immunostaining is commonly used for the discrimination of urothelial transitional cell carcinoma in-situ from non-neoplastic changes in the urothelium.
ELISA: order BSA-free format for coating Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1 ug/ml WB: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the HCAM antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 156-3C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Baboon, Green Monkey