Specifications
- Antibody Type:Primary
- Antigen Name:CD6 molecule
- Antigen Symbol:CD6
- Clonality:Monoclonal
- Clone:3F7B5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P30203
- Antigen Synonyms:CD6
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:The human rheumatoid synovial T cell line ST-1 was used as the immunogen for this CD6 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76193-832
Specifications
About this item
CD6 is a type I transmembrane glycoprotein that contains a 24-amino acid signal sequence, three extracellular “scavenger receptor cysteine-rich” (SRCR) domains, a membrane-spanning domain and a 44-amino acid cytoplasmic domain. The glycoprotein is tyrosine phosphorylated during TCR-mediated T cell activation. It shows significant homology to CD5. CD6 is present on mature thymocytes, peripheral T cells and a subset of B cells. CD6 antibody can be used to deplete T cells from bone marrow transplants to prevent graft versus host disease.
Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate Buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 3F7B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human