- Antibody Type:Primary
- Antigen Name:CD3e molecule, epsilon (CD3-TCR complex)
- Antigen Symbol:CD3E
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Bovine,Guinea pig,Dog,Rat,Mouse,Elephant,Cat,Primates,Hamster,Opossum,Frog,Pig,Rabbit,Horse,Mole rat,Sheep,Chicken,Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P07766
- Antigen Synonyms:CD3E
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Amino acids ERPPPVPNPDYEP (156-168) were used as the immunogen for this CD3e antibody.
- Purification:Protein A purified CD3e antibody
- Cat. No.:76193-826
This antibody recognizes the epsilon-chain of CD3 (CD3e), which consists of five different polypeptide chains (designated as gamma, delta, epsilon, zeta, and eta) with MW ranging from 16-28kDa. The CD3 complex is closely associated at the lymphocyte cell surface with the Tcell antigen receptor (TCR). Reportedly, CD3 complex is involved in signal transduction to the Tcell interior following antigen recognition. The CD3 antigen is first detectable in early thymocytes and probably represents one of the earliest signs of commitment to the Tcell lineage. In cortical thymocytes, CD3 is predominantly intra-cytoplasmic. However, in medullary thymocytes, it appears on the Tcell surface. CD3 antigen is a highly specific marker for Tcells, and is present in majority of Tcell neoplasms.
Flow Cytometry: use the CD3e antibody at 0.5-1 ug/million cells WB: 0.5-1.0 ug/ml IP: 1-2ug/500ug protein lysate The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the CD3e antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: CD3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Primate, Guinea Pig, Hamster, Horse, Mole Rat, Pig, Rabbit, Sheep, Cat, Chicken, Bovine, Dog, Frog, Opossum, Elephant