Specifications
- Antibody Type:Primary
- Antigen Name:complement component 4A (Rodgers blood group)
- Antigen Symbol:C4A
- Clonality:Monoclonal
- Clone:C4D204
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P0C0L4
- Antigen Synonyms:C4A
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Human protein was used as the immunogen for this C4d antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-430
Specifications
About this item
This antibody is specific to Complement 4d (C4d) and it reacts with the secreted as well as cell-bound protein. C4d is a degradation product of the activated complement factor C4b. Complement 4b is typically activated by binding of antibodies to specific target molecules. Following activation and degradation of the C4 molecule, thio-ester groups are exposed, which allow transient, covalent binding of the degradation product C4d to endothelial cell surfaces and extracellular matrix components of vascular basement membranes near the sites of C4 activation. The presence of C4d in peritubular capillaries is a key indicator for acute humoral (i.e. antibody-mediated) rejection of kidney, heart, pancreas and lung allografts. As an established marker of antibody-mediated acute renal allograft rejection and its proclivity for endothelium, this component can be detected in peritubular capillaries in chronic renal allograft rejection as well as hyperacute rejection, acute vascular rejection, acute cellular rejection, and borderline rejection. C4d has been shown to be a significant predictor of transplant kidney graft survival. C4d antibody, combined with antibody to C3d, can be utilized as a tool for diagnosis of allograft rejection that may warrant a prompt and aggressive anti-rejection treatment.
IF: 1:50-1:100 IHC (FFPE): 1:200-1:400 for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) ELISA: 1-2 ug/ml; order the BSA-free format The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the C4d antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 1mM EDTA, pH 7.5-8.5, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: C4A
Clonality: Monoclonal
Clone: C4D204
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human