ContactUsLinkComponent
TransIT®-VirusGEN™ Transfection Reagent, Mirus Bio
TransIT®-VirusGEN™ Transfection Reagent, Mirus Bio
Supplier:  Mirus Bio
Certificates
undefined
TransIT®-VirusGEN™ Transfection Reagent, Mirus Bio
Supplier:  Mirus Bio
Specifications
  • Bioproduction:
    Yes
  • Cell Specific:
    293
  • CRISPR/Cas 9:
    Yes
  • DNA:
    Yes
  • Virus production:
    Yes
Specifications
TransIT-VirusGEN SELECT Transfection Reagent
Product Information