SEAP Assay Kit, OZ Biosciences
Catalog # 103258-312
Supplier: OZ Biosciences
CAS Number:
Specifications
- Description:Assay Kit
- Brand Name:OZBI
- Cat. No.:103258-312
- Buffer volume:Reaction buffer #1 component B: 125 ml
Reaction buffer #2 component C: 125 μl
Dilution buffer component D: 50 ml - No. of assays/kit:500 assays
- Volume:pNPP - containing assay buffer component A: 10× 1 ml
- Supplier no.:SP00500
Specifications
About this item
OZ Biosciences SEAP Assay Kit (Secreted Alkaline Phosphatase) is a colorimetric assay for sensitive quantification of SEAP in culture medium from transfected cells or tissues
- Designed to detect SEAP reporter gene expression
- Fast and easy analysis
- Colorimetric-measure with a standard spectrophotometer or an ELISA reader (405 nm)
- Multiple samples can be analyzed in low volumes
- Convenient and economical packaging: no tablet, no powder
Alkaline phosphatase cleaves the colourless 4-nitrophenylphosphate (pNPP) substrate to give a yellow p-nitrophenolate product in alkaline solutions. It is commonly used as a reporter gene expression and can be accurately quantified. It is a truncated form of placental alkaline phosphatase (GPI-anchored protein).
Delivery: Shipped at room temperature
Packaging: Store at +4°C