Order Entry
Puerto Rico
ContactUsLinkComponent
Native R-phycoerythrin (from Algae)
Native R-phycoerythrin (from Algae)
Catalog # 103258-164
Supplier:  OZ Biosciences
CAS Number:  
undefined
Native R-phycoerythrin (from Algae)
Catalog # 103258-164
Supplier:  OZ Biosciences
Supplier Number:  PHY0100
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Native
  • Source:
    Algae
  • Size:
    100 µl
  • Protein Synonyms:
    R-PE
  • Protein/Peptide Name:
    R-phycoerythrin
  • Cat. No.:
    103258-164
  • Supplier no.:
    PHY0100

Specifications

About this item

OZ Biosciences R-phycoerythrin (R-PE) is a naturally fluorescent protein from algae and is composed of a 240KD protein covalently linked to a number of fluorophores called phycobilins.

  • Absorption maximum: 498/565 nm
  • Emission maximum: 573 nm
  • Extinction coefficient: 1.96x10^6 M-1 cm-1
  • Quantum yield - 0,84

Transfection Control

Delivery: Shipped at room temperature
Packaging: Store at +4°C and protect from the light