Order Entry
Puerto Rico
ContactUsLinkComponent
TBE 10X Buffer, MP Biomedicals
TBE 10X Buffer, MP Biomedicals
Catalog # 76180-758
Supplier:  MP Biomedicals
CAS Number:  
undefined
TBE 10X Buffer, MP Biomedicals
Catalog # 76180-758
Supplier:  MP Biomedicals
Supplier Number:  11TBE10X02
CAS Number:  

Specifications

  • Description:
    TBE 10X Buffer
  • Size:
    1L
  • Cat. No.:
    76180-758

Specifications

About this item

TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis, after dilution to working concentrations.

  • Use with non-denaturing or denaturing gels
  • pH: 8.2 - 8.4 (Lit.)
  • Soluble in water

TBE buffer maintains the structural integrity of nucleic acids and more suitable for their size analysis. TBE has a greater buffering capacity and will give sharper resolution. TBE is better suited for high-voltage (>150 V) electrophoresis because of its higher buffering capacity.

TBE is routinely used for DNA automated sequencing gel. TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids