Order Entry
Puerto Rico
ContactUsLinkComponent
TAE 50X Buffer, MP Biomedicals
TAE 50X Buffer, MP Biomedicals
Catalog # 76180-756
Supplier:  MP Biomedicals
CAS Number:  
undefined
TAE 50X Buffer, MP Biomedicals
Catalog # 76180-756
Supplier:  MP Biomedicals
Supplier Number:  11TAE50X01
CAS Number:  

Specifications

  • Description:
    TAE 50X Buffer
  • Size:
    1L
  • Cat. No.:
    76180-756

Specifications

About this item

Buffer for Electrophoresis.

  • Used to prepare buffers for polyacrilamide and Agarose gel electrophoresis.
  • Components: Tris Base 2M; Acetic acid 1M; EDTA 0,05 M
  • Filtered through 0.2pm PTFE aseptically filled
  • Conductivity of 1X conc. (25°C): 1.3 mS/cm Conductivity of 50X conc. (25°C): 18 mS/cm

TAE running buffer is the most commonly used buffer for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis.