Order Entry
Puerto Rico
ContactUsLinkComponent
SSC 20X Buffer, MP Biomedicals
SSC 20X Buffer, MP Biomedicals
Catalog # 76180-744
Supplier:  MP Biomedicals
CAS Number:  
SSC 20X Buffer, MP Biomedicals
Catalog # 76180-744
Supplier:  MP Biomedicals
Supplier Number:  11RIST2050
CAS Number:  

Specifications

  • Description:
    SSC 20X Buffer
  • Size:
    500mL
  • Cat. No.:
    76180-744

Specifications

About this item

SSC Buffer used in electrophoresis and in sedimentation studies of DNA-histone aggregates.

  • Molecular Biology Grade
  • Clear, Colorless Solution
  • 20x Concentrated
  • Formulation: 3 M Trisodium citrate
  • Sterility: 0.2 μm filtered

SSC is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.