Order Entry
Puerto Rico
ContactUsLinkComponent
Phosphate Buffered Saline Tablets (PBS), MP Biomedical, MP Biomedicals
Phosphate Buffered Saline Tablets (PBS), MP Biomedical, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
Phosphate Buffered Saline Tablets (PBS), MP Biomedical, MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • Sterility:
    Autoclave for 1 minute at 115 °C
  • Storage Temperature:
    Store at Room Temperature(15…30° C); Desiccate

Specifications

Related Information

Frequently Bought Together