Order Entry
Puerto Rico
ContactUsLinkComponent
1X Hanks' Balanced Salt Solution (Modified) without Calcium, Magnesium, Phenol Red, MP Biomedicals
1X Hanks' Balanced Salt Solution (Modified) without Calcium, Magnesium, Phenol Red, MP Biomedicals
Catalog # 76180-680
Supplier:  MP Biomedicals
CAS Number:  
1X Hanks' Balanced Salt Solution (Modified) without Calcium, Magnesium, Phenol Red, MP Biomedicals
Catalog # 76180-680
Supplier:  MP Biomedicals
Supplier Number:  091810554
CAS Number:  

Specifications

  • Color:
    Clear
  • Description:
    1X Hanks' Balanced Salt Solution (Modified) without Ca, Mg, Phenol Red
  • Size:
    500mL
  • Cat. No.:
    76180-680

Specifications

About this item

Used to minimize cell attachment or aggregation for growth of cells at low densities

  • Sterile
  • Suitable for Cell Culture
  • Formulation: Inorganic Salts: Potassium Chloride [KCl] 400.00 mg/liter, Potassium Phosphate Monobasic [KH2PO4] 60.00 mg/liter, Sodium Bicarbonate [NaHCO3] 350.00 mg/liter, Sodium Chloride [NaCl] 8000.00 mg/liter, Sodium Phosphate Dibasic [Na2HPO4] 47.50 mg/liter; Other Dextrose: 1000.00 mg/liter
  • pH 6.9-7.4
  • Osmolality: 258-298 mOsm
  • Store at +4 °C

Calcium and magnesium support cell adhesion. Hanks' Balanced Salt Solution (Modified) is used to minimize cell attachment or aggregation. Phenol red has been shown to interfere with the growth of some cells at cloning densities. Use this medium when growing cells at low densities.