Hanks' Balanced Salts (Modified), 1X, without Magnesium and Calcium, MP Biomedicals
Catalog # 76180-678
Supplier: MP Biomedicals
CAS Number:
Specifications
- Color:Red
- Description:1X Hanks' Balanced Salts (Modified), without Mg and Ca
- Size:500mL
- Cat. No.:76180-678
Specifications
About this item
Used to minimize cell attachment or aggregation.
- Sterile
- Suitable for Cell Culture
- Formulation: Inorganic Salts: Potassium Chloride [KCl] 400.00000 mg/L, Potassium Phosphate Monobasic [KH2PO4] 60.00000 mg/L, Sodium Bicarbonate [NaHCO3] 350.00000 mg/L, Sodium Chloride [NaCl] 8000.00000 mg/L, Sodium Phosphate Dibasic [Na2HPO4] 47.50000 mg/L, Dextrose 1000.00000 mg/L, Phenol Red Sodium Salt 17.00000 mg/L
- pH: 6.9-7.4
- Osmolality 258 - 298 mM/kg
Calcium and magnesium support cell adhesion. Hanks' Balanced Salts (Modified), without Magnesium and Calcium, is used to minimize cell attachment or aggregation.