Order Entry
Puerto Rico
ContactUsLinkComponent
Tris-Borate-EDTA (TBE) Buffer, MP Biomedicals
Tris-Borate-EDTA (TBE) Buffer, MP Biomedicals
Catalog # 76180-658
Supplier:  MP Biomedicals
CAS Number:  
Tris-Borate-EDTA (TBE) Buffer, MP Biomedicals
Catalog # 76180-658
Supplier:  MP Biomedicals
Supplier Number:  04816721
CAS Number:  

Specifications

  • Description:
    Tris-Borate-EDTA (TBE) Buffer
  • Size:
    1L
  • Cat. No.:
    76180-658

Specifications

About this item

TBE buffer is a solution of 1M Tris, 0.9M boric acid and 0.01M EDTA disodium salt, prepared from MP's Ultra Pure products.

  • Suitable for electrophoresis
  • Clear, Colorless Solution
  • pH (as is ): 8.3 - 8.5, pH (1X): 8.23 - 8.43
  • Store at Room Temperature(15-30 °C)

TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel. TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids. It is used to prepare a 1 X buffer for polyacrylamide and agarose gel electrophoresis. When diluted to a 1X concentration, it yields a TBE buffer solution that contains 100 mM Tris, 90 mM boric acid and 1mM EDTA. Applied voltages of less than 5 V/cm (distance between the electrodes of the unit) are recommended for maximum resolution.