Twin-Cartridge Adapter for Ultravue and Ultra Elite Facepieces, MSA
Catalog # 300081-118
Supplier: Safety & Industrial Supplies
Specifications
- Description:Twin-Cartridge Adapter, Plastic
- Environmentally Preferable:
- Cat. No.:300081-118
- Supplier no.:454-803622
Specifications
About this item
Easy to install adapter is designed for converting Ultra Elite or Ultravue facepiece to a twin cartridge respirator.