ContactUsLinkComponent
Mesenchymal Stem Cell Medium, iXCells Biotechnologies
Mesenchymal Stem Cell Medium, iXCells Biotechnologies
  103521-938
 :  
Mesenchymal Stem Cell Medium, iXCells Biotechnologies
  103521-938
 :  MD-0037-500ML
 :  
  • Media Format:
    Liquid
  • Size:
    500 ml
  • Cat. No.:
    103521-938
Mesenchymal Stem Cell Medium is designed to promote mesenchymal stem cells proliferation in culture
This medium is fully supplemented with fetal bovine serum, growth factors and antibiotic. This medium is sterile filtered and is ready for use.