Fibroblast Growth Medium, iXCells Biotechnologies
Catalog # 103521-470
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Media Format:Liquid
- Size:500 ml
- Cat. No.:103521-470
Specifications
About this item
Fibroblast Growth Medium is designed to promote fibroblast proliferation in culture
This medium is fully supplemented with fetal bovine serum and growth factors. This medium is sterile filtered and is ready for use.