Adipocyte Differentiation Medium, iXCells Biotechnologies
Catalog # 103521-464
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Media Format:Liquid
- Size:250 ml
- Cat. No.:103521-464
Specifications
About this item
Adipocyte Differentiation Medium designed to efficiently induce preadipocytes or adipose-derived stem cells (ADSCs) differentiation into adipocytes.
- Optimized for efficient adipocyte differentiation
- Medium is fully supplemented and containing 10% fetal bovine serum
- Read-to-use