Mouse Aortic Endothelial Cells (MAOEC), iXCells Biotechnologies
Catalog # 103521-246
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Size:0.5 million cells/vial
- Cat. No.:103521-246
Specifications
About this item
Mouse Aortic Endothelial Cells (MAOEC); Frozen
- High-quality Primary Mouse Aortic Endothelial Cells (MAOEC)
- High purity
MAOEC are isolated from aorta of C57BL6 mice. MAOEC are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast and fungi.