Rat Lung Fibroblasts - adult (RLuFB-a), iXCells Biotechnologies
Catalog # 103521-240
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Size:0.5 million cells/vial
- Cat. No.:103521-240
Specifications
About this item
Rat Lung Fibroblasts - adult (RLuFB-a); Frozen
- High-quality Primary Rat Lung Fibroblasts - adult (RLuFB-a)
- High purity
RLuFB-a are isolated from rat lung tissue. RLuFB-a are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi.