Vydac 302IC, Anion Exclusion HPLC Column
Supplier: Avantor
Certificates
About this item
Vydac 302IC is a low capacity anion-exchange (quaternary amine) material based on a high purity 10 µm large pore size silica.
- Used with standard HPLC systems for analysis of common ions
- Widely used for environmental analysis
This column is well known for 'non-suppressed' ion chromatography applications.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Catalog No:
- 75870-424
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental Certification:
- ISO 14001:2015 (530840 UM15)