Human Peripheral Blood CD19+ B Cells, iXCells Biotechnologies
Catalog # 103521-902
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Size:5 million cells/vial
- Cat. No.:103521-902
Specifications
About this item
Human Peripheral Blood CD19+ B Cells; Frozen
- High-quality Human Peripheral Blood CD19+ B Cells
- High purity
- IRB-approved
Human Peripheral Blood CD19+ B Cells are isolated from human peripheral blood mononuclear cells (PBMCs) using positive immunomagnetic selection. > 95% of the cells are CD19 positive.