Human Dermal Microvascular Endothelial Cells (HDMVEC), iXCells Biotechnologies
Catalog # 103521-202
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Size:0.5 million cells/vial
- Cat. No.:103521-202
Specifications
About this item
Human Dermal Microvascular Endothelial Cells (HDMVEC) isolated from blood vessels of skin.
- High-quality primary Human Dermal Microvascular Endothelial Cells
- High purity
- IRB-approved
Human Dermal Microvascular Endothelial Cells (HDMVEC); Frozen; 0.5 million cells/vial. HDMVEC are isolated from blood vessels of skin. HDMVEC are characterized by CD31 and vWF/Factor VIII staining. HDMVEC are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi.