Human iPSC-Derived Neural Stem Cells, iXCells Biotechnologies
103521-784
:
- Cell Format:Cryopreserved
- Size:2.0 million cells/vial
- Cat. No.:103521-784
Human iPSC-derived neural stem cells; iXCells™ hiPSC-derived neural stem cells express typical markers of neural stem and progenitor cells, e.g
- High-quality human neural stem cells derived from iPSCs
- High purity
- IRB-approved
Nestin, Pax6 and Sox1, with the purity higher than 97%. Negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi.