Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 490 HIV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® 490 HIV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-148
Supplier:  Anaspec
CAS Number:  
SensoLyte® 490 HIV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-148
Supplier:  Anaspec
Supplier Number:  AS-71127
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 Assays (96-well plate) or 1250 Assays (384-well)
  • Cat. No.:
    103010-148
  • Storage temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-71127

Specifications

About this item

HIV protease (PR) is identified as an important drug-screening target for the design of selective acquired immunodeficiency syndrome (AIDS) therapeutics.

The SensoLyte® 490 HIV Protease Assay Kit uses an optimized FRET peptide substrate for a continuous measurement of HIV protease activities. This FRET-based fluorogenic substrate is derived from a natural processing site for HIV-1 PR. Incubation of the recombinant HIV-1 PR with this substrate results in specific cleavage and a time-dependent increase in fluorescence intensity that is linearly related to the extent of substrate hydrolysis. Because of its simplicity and precision in the determination of reaction rates required for kinetic analysis, this kit offers many advantages over the commonly used HPLC or electrophoresis-based assays for peptide substrate hydrolysis by retroviral PRs.