Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 520 HCV Protease Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 HCV Protease Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-174
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 HCV Protease Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-174
Supplier:  Anaspec
Supplier Number:  AS-71145
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays (96-well plate) or 300 Assays (384-well)
  • Cat. No.:
    103010-174
  • Storage temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-71145

Specifications

About this item

The SensoLyte® 520 HCV Assay Kit uses a new FRET peptide substrate that incorporates 5-FAM (fluorophore) and QXL® 520 (quencher) for a continuous measurement of enzyme activities. In the intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon cleavage of the FRET peptide by HCV NS3/4a protease, the fluorescence of 5-FAM is recovered and can be continuously monitored at excitation/emission = 490 nm/520 nm. With superior fluorescence quantum yield and longer emission wavelength, this 5-FAM/QXL® 520-based FRET peptide shows less interference from autofluorescence of test compounds and cellular components. This assay is ten times more sensitive than an EDANS/DABCYL based assay and can detect 0.1 pmole of HCV NS3/4a protease. The assays are performed in a convenient 96-well or 384-well microplate format.