Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® AFC Thrombin Activity Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® AFC Thrombin Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-468
Supplier:  Anaspec
CAS Number:  
SensoLyte® AFC Thrombin Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-468
Supplier:  Anaspec
Supplier Number:  AS-72130
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-468
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72130

Specifications

About this item

Thrombin, a serine protease, plays a central role in hemostasis by converting soluble plasma fibrinogen into an insoluble fibrin clot and by promoting platelet aggregation

The SensoLyte® AFC Thrombin Assay Kit is optimized for screening of enzyme inhibitors. This kit contains a fluorogenic substrate which shows high reactivity and low background. Thrombin cleaves the substrate resulting in the release of the AFC (7-amido-4-trifluoromethylcoumarin) fluorophore. Fluorescence can be monitored at excitation/emission= 380 nm/500 nm. The assays are performed in a convenient 96-well or 384-well microplate format.

Thrombin also influences a number of normal and pathological processes, including inflammation, tissue repair, embryogenesis, angiogenesis, and tumor invasion.