Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 520 Thrombin Activity Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 Thrombin Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-466
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 Thrombin Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-466
Supplier:  Anaspec
Supplier Number:  AS-72129
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-466
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72129

Specifications

About this item

Thrombin, a serine protease, plays a central role in hemostasis by converting soluble plasma fibrinogen into an insoluble fibrin clot and by promoting platelet aggregation

The SensoLyte® 520 Thrombin Assay Kit is optimized for detecting thrombin activity. This kit contains a novel internally quenched 5-FAM/QXL® 520 FRET substrate for thrombin. Thrombin cleaves the FRET substrate into two separate fragments resulting in the release of 5-FAM fluorescence which can be monitored at excitation/emission= 490 nm/520 nm. The long wavelength fluorescence of 5-FAM is less interfered by the autofluorescence of components in biological samples and test compounds. The assay can be performed in a convenient 96-well or 384-well microplate format for screening of enzyme inhibitors.

Thrombin also influences a number of normal and pathological processes, including inflammation, tissue repair, embryogenesis, angiogenesis, and tumor invasion.