Specifications
- Size:500 Assays
- Cat. No.:103010-480
- Storage temperature:– 20 °C
- Includes:The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
- Supplier no.:AS-72138
Specifications
About this item
Thiols, compounds that contain sulfhydryl group, are powerful reducing agents capable of acting as antioxidants, enzyme cofactors, and neuromodulators.
The SensoLyte® 520 Thiol Quantitation Assay Kit provides a convenient, highly sensitive fluorescent assay for measurement of thiol concentrations. The novel reagent supplied in the kit, upon reaction with thiols, generates fluorescence which can be easily detected at excitation/emission=490/520 nm. The assay has a linear range of 0.04 to 160 µM thiol.