Order Entry
Puerto Rico
ContactUsLinkComponent
 
SensoLyte® 520 FRET SIRT2 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-588
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® 520 FRET SIRT2 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-588
Supplier:  Anaspec Inc
Supplier Number:  AS-72189
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-588
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72189

Specifications

About this item

Histone deacetylase (HDAC) enzymes act as transcriptional repressors of genes through the deacetylation of lysine residues on histone proteins

The SensoLyte® 520 FRET SIRT2 Assay Kit provides a convenient, two-step homogeneous procedure for measuring sirtuin 2 activity using FRET substrate. In the first step, an acetylated substrate is incubated with SIRT2 containing samples. Deacetylation of substrate first sensitizes it, so that, in the second step mixing with the SIRT developer generates a fluorophore, which can then be detected at Ex/Em= 490/520 nm. This kit can be used for high throughput screening of SIRT2 inhibitors/activators. The long wavelength fluorescence of FRET substrate is less interfered by the autofluorescence of cell components and test compounds. Assays are performed in a standard 96-well plate format.

Sirtuins (SIRTs) are a unique class of deacetylases (class III HDACs) that require NAD+ as a cofactor. Sirtuin 2 (SIRT2) is a cytoplasmic protein involved in regulation of cell cycle, DNA repair and tumorogenesis.