Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® Red Protease Assay Kit (Fluorimetric), AnaSpec
SensoLyte® Red Protease Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-168
Supplier:  Anaspec
CAS Number:  
SensoLyte® Red Protease Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-168
Supplier:  Anaspec
Supplier Number:  AS-71140
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 Assays
  • Cat. No.:
    103010-168
  • Storage temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-71140

Specifications

About this item

The SensoLyte® Red Protease Assay Kit is widely used for continuous detection of generic protease activities. The kit uses a casein derivative that is heavily labeled with a rhodamine derivative, resulting in almost total quenching of the conjugate's fluorescence. Compared to the SensoLyte® Green Protease Assay Kit, this kit contains a red fluorescent casein derivative. Protease-catalyzed hydrolysis relieves this quenching conjugate, yielding brightly red fluorescent dye-labeled peptides (Ex/Em=546/575 nm). The increase in fluorescence intensity is directly proportional to protease activity. The SensoLyte® protease assay kits do not require any separation steps and can be used to continuously measure the kinetics of a variety of exopeptidases and endopeptidases.