Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® Luminescent Peroxidase Assay Kit (Luminometric), AnaSpec Inc.
SensoLyte® Luminescent Peroxidase Assay Kit (Luminometric), AnaSpec Inc.
Catalog # 103010-462
Supplier:  Anaspec
CAS Number:  
SensoLyte® Luminescent Peroxidase Assay Kit (Luminometric), AnaSpec Inc.
Catalog # 103010-462
Supplier:  Anaspec
Supplier Number:  AS-72127
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 Assays
  • Cat. No.:
    103010-462
  • Storage temperature:
    +4 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72127

Specifications

About this item

Peroxidases catalyze oxidation-reduction reactions and play an important role in protecting cell from oxidative injury

The SensoLyte® Luminescent Peroxidase Assay Kit provides highly sensitive chemiluminescent substrate to quantify peroxidase activity in solutions, in cell extracts, in live cells. The kit contains: chemiluminescent substrate, positive control (horseradish peroxidase), assay buffer, and an optimized ‘mix and read’ assay protocol that is compatible with HTS liquid handling instruments.

HRP-protein conjugates are extensively used as secondary detection reagents in ELISA, immunohistochemistry and assay development.